Product Details

SNP ID
rs200121663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52705077 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTTGACAGGAGTGAATTACGCA[C/T]GAAAGCCTTGTCACAAATTGTACAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF611 PubMed Links

Gene Details

Gene
ZNF611
Gene Name
zinc finger protein 611
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161499.1 Intron NP_001154971.1
NM_001161500.1 Intron NP_001154972.1
NM_001161501.1 Intron NP_001154973.1
NM_030972.3 Intron NP_112234.3

View Full Product Details