Product Details

SNP ID
rs202026383
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35250421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACCAGATGACCTCGACCCCCAC[A/G]CAACCCATCGTCATCTGGAAGTACA
Phenotype
MIM: 616582
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LSR PubMed Links

Gene Details

Gene
LSR
Gene Name
lipolysis stimulated lipoprotein receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001260489.1 583 Silent Mutation ACA,ACG T120T NP_001247418.1
NM_001260490.1 583 Silent Mutation ACA,ACG T120T NP_001247419.1
NM_015925.6 583 Silent Mutation ACA,ACG T120T NP_057009.3
NM_205834.3 583 Silent Mutation ACA,ACG T120T NP_991403.1
NM_205835.3 583 Silent Mutation ACA,ACG T120T NP_991404.1
XM_005258980.1 583 Silent Mutation ACA,ACG T120T XP_005259037.1
XM_005258982.1 583 Silent Mutation ACA,ACG T120T XP_005259039.1
XM_011527026.1 583 Silent Mutation ACA,ACG T120T XP_011525328.1

View Full Product Details