Product Details

SNP ID
rs200503204
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17942822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGACACGCTGCGCCGAGACCATC[A/G]GCTCAGGGAGGCCCCGGACACAGGT
Phenotype
MIM: 602982
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC124 PubMed Links

Gene Details

Gene
CCDC124
Gene Name
coiled-coil domain containing 124
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136203.1 371 Missense Mutation CAG,CGG Q109R NP_001129675.1
NM_138442.3 371 Missense Mutation CAG,CGG Q109R NP_612451.1
Gene
KCNN1
Gene Name
potassium calcium-activated channel subfamily N member 1
There are no transcripts associated with this gene.

View Full Product Details