Product Details

SNP ID
rs200574015
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4013170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACCATGCACTCCACCTACGACC[A/G]GGCCGGCGCTGTGCCCAGGACTCCG
Phenotype
MIM: 605989
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PIAS4 PubMed Links

Gene Details

Gene
PIAS4
Gene Name
protein inhibitor of activated STAT 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015897.3 438 Missense Mutation CAG,CGG Q92R NP_056981.2
XM_011528060.2 438 Missense Mutation CAG,CGG Q111R XP_011526362.1
XM_017026868.1 438 UTR 5 XP_016882357.1

View Full Product Details