Product Details

SNP ID
rs200679414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19540374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCTGCCGTCCGCCGTCGGCGAG[C/T]GCGTGCACTTGAACCCCACGCGCGG
Phenotype
MIM: 612419
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CILP2 PubMed Links

Gene Details

Gene
CILP2
Gene Name
cartilage intermediate layer protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153221.2 419 Missense Mutation CGC,TGC R112C NP_694953.2
Gene
YJEFN3
Gene Name
YjeF N-terminal domain containing 3
There are no transcripts associated with this gene.

View Full Product Details