Product Details

SNP ID
rs200747180
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7928088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGAGGTGCTTACGTCGACGTTGT[C/T]AATGTCTAGGATGCGAGAATGGAAC
Phenotype
MIM: 600135 MIM: 605076 MIM: 605058
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CTXN1 PubMed Links
Additional Information
For this assay, SNP(s) [rs11542187] are located under a probe and SNP(s) [rs34425383] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CTXN1
Gene Name
cortexin 1
There are no transcripts associated with this gene.

Gene
SNAPC2
Gene Name
small nuclear RNA activating complex polypeptide 2
There are no transcripts associated with this gene.

Gene
TGFBR3L
Gene Name
transforming growth factor beta receptor 3 like
There are no transcripts associated with this gene.

Gene
TIMM44
Gene Name
translocase of inner mitochondrial membrane 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006351.3 1289 Missense Mutation AAC,GAC N373D NP_006342.2

View Full Product Details