Product Details

SNP ID
rs201781055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15949493 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTCATGGGGATGATGGTGACAAC[A/G]ATAGTTTTCCACCTCACTTTCTGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10H4 PubMed Links

Gene Details

Gene
OR10H4
Gene Name
olfactory receptor family 10 subfamily H member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004465.1 486 Silent Mutation ACA,ACG T162T NP_001004465.1

View Full Product Details