Product Details

SNP ID
rs202096937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11487066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCCTCCCCGTCCGCCTCTGCCT[C/T]AGGCTCTGCGCTCTCAGGCACCGTT
Phenotype
MIM: 603458 MIM: 611371
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELAVL3 PubMed Links

Gene Details

Gene
ELAVL3
Gene Name
ELAV like neuron-specific RNA binding protein 3
There are no transcripts associated with this gene.

Gene
MIR7974
Gene Name
microRNA 7974
There are no transcripts associated with this gene.

Gene
ZNF653
Gene Name
zinc finger protein 653
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138783.3 1401 Missense Mutation AAG,GAG K422E NP_620138.2

View Full Product Details