Product Details

SNP ID
rs200474061
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1881302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCCGCGGGCGCTCTTGGTGGGGA[A/T]GACCTCGATGGTGTCCAGCTCGCGC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ABHD17A PubMed Links

Gene Details

Gene
ABHD17A
Gene Name
abhydrolase domain containing 17A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130111.1 3543 Intron NP_001123583.1
NM_031213.3 3543 Intron NP_112490.3
XM_011528338.1 3543 Missense Mutation ATC,TTC I89F XP_011526640.1
XM_017027341.1 3543 Missense Mutation ATC,TTC I89F XP_016882830.1
XM_017027342.1 3543 UTR 5 XP_016882831.1

View Full Product Details