Product Details

SNP ID
rs202033824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3578005 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACGGGCGAAACGCCCACGCTGG[A/G]CACTCTGGACTTCTACATGGCCCGG
Phenotype
MIM: 608792 MIM: 605535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GIPC3 PubMed Links

Gene Details

Gene
GIPC3
Gene Name
GIPC PDZ domain containing family member 3
There are no transcripts associated with this gene.

Gene
HMG20B
Gene Name
high mobility group 20B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006339.2 901 Missense Mutation GAC,GGC D278G NP_006330.2
XM_017026144.1 901 Missense Mutation GAC,GGC D277G XP_016881633.1

View Full Product Details