Product Details

SNP ID
rs200510680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39453361 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTGCCTGACCCCTGTAGGTAGA[C/T]GAAGAGCGGCGGAGTGCAGCGGGCA
Phenotype
MIM: 602102
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SUPT5H PubMed Links

Gene Details

Gene
SUPT5H
Gene Name
SPT5 homolog, DSIF elongation factor subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111020.2 182 Silent Mutation GAC,GAT D27D NP_001104490.1
NM_001130824.1 182 Silent Mutation GAC,GAT D27D NP_001124296.1
NM_001130825.1 182 Silent Mutation GAC,GAT D27D NP_001124297.1
NM_001319990.1 182 Silent Mutation GAC,GAT D27D NP_001306919.1
NM_001319991.1 182 Silent Mutation GAC,GAT D27D NP_001306920.1
NM_003169.3 182 Silent Mutation GAC,GAT D27D NP_003160.2
XM_017027174.1 182 Intron XP_016882663.1

View Full Product Details