Product Details

SNP ID
rs200511054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41848851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGGAAGCCTCGCCCTTGTCCTG[G/T]ACTCCGGTGCCTCCTGGCCCTTGGG
Phenotype
MIM: 614806
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DMRTC2 PubMed Links

Gene Details

Gene
DMRTC2
Gene Name
DMRT like family C2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040283.2 609 Missense Mutation TGG,TGT W168C NP_001035373.1
XM_011527214.1 609 Missense Mutation TGG,TGT W45C XP_011525516.1
XM_011527215.1 609 Missense Mutation TGG,TGT W45C XP_011525517.1
XM_011527216.2 609 Missense Mutation GAC,TAC D187Y XP_011525518.1
XM_017027123.1 609 Missense Mutation TGG,TGT W173C XP_016882612.1
XM_017027124.1 609 Missense Mutation TGG,TGT W171C XP_016882613.1
XM_017027125.1 609 Missense Mutation TGG,TGT W168C XP_016882614.1
XM_017027126.1 609 Missense Mutation TGG,TGT W168C XP_016882615.1
XM_017027127.1 609 Missense Mutation TGG,TGT W168C XP_016882616.1
XM_017027128.1 609 Missense Mutation TGG,TGT W168C XP_016882617.1
XM_017027129.1 609 Missense Mutation TGG,TGT W173C XP_016882618.1
Gene
LYPD4
Gene Name
LY6/PLAUR domain containing 4
There are no transcripts associated with this gene.

View Full Product Details