Product Details

SNP ID
rs200615699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45713331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCAGTGGCCAAGGCGGGTGGTGT[A/G]TTCTCTGAGTGGGCAGGCCCATGGT
Phenotype
MIM: 609117
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO46 PubMed Links

Gene Details

Gene
FBXO46
Gene Name
F-box protein 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080469.1 288 Silent Mutation AAC,AAT N55N NP_001073938.1
Gene
QPCTL
Gene Name
glutaminyl-peptide cyclotransferase like
There are no transcripts associated with this gene.

View Full Product Details