Product Details

SNP ID
rs201648976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54888039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAAACCCTTCCTCTCTGCAGAT[C/T]GGGGTCTGGTGTTGATGCCAGGAGA
Phenotype
MIM: 147045
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FCAR PubMed Links

Gene Details

Gene
FCAR
Gene Name
Fc fragment of IgA receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002000.3 555 Missense Mutation CGG,TGG R132W NP_001991.1
NM_133269.3 555 Missense Mutation CGG,TGG R132W NP_579803.1
NM_133271.3 555 Intron NP_579805.1
NM_133272.3 555 Missense Mutation CGG,TGG R120W NP_579806.1
NM_133273.3 555 Intron NP_579807.1
NM_133274.3 555 Missense Mutation CGG,TGG R120W NP_579808.1
NM_133277.3 555 Missense Mutation CGG,TGG R23W NP_579811.1
NM_133278.3 555 Missense Mutation CGG,TGG R120W NP_579812.1
XM_011526625.2 555 Missense Mutation CGG,TGG R105W XP_011524927.1
XM_017026473.1 555 Missense Mutation CGG,TGG R105W XP_016881962.1
XM_017026474.1 555 Missense Mutation CGG,TGG R105W XP_016881963.1

View Full Product Details