Product Details

SNP ID
rs201716955
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11576557 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACACATTGGTCTGTGGGATCTTGA[A/C]GTGCAGGCGGTAGAAAGGGCTGGGG
Phenotype
MIM: 171640 MIM: 612248
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACP5 PubMed Links

Gene Details

Gene
ACP5
Gene Name
acid phosphatase 5, tartrate resistant
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111034.2 824 Missense Mutation GTC,TTC V141F NP_001104504.1
NM_001111035.2 824 Missense Mutation GTC,TTC V141F NP_001104505.1
NM_001111036.2 824 Missense Mutation GTC,TTC V141F NP_001104506.1
NM_001322023.1 824 Missense Mutation GTC,TTC V141F NP_001308952.1
NM_001611.4 824 Missense Mutation GTC,TTC V141F NP_001602.1
XM_005259938.1 824 Missense Mutation GTC,TTC V141F XP_005259995.1
XM_011528069.2 824 Missense Mutation GTC,TTC V141F XP_011526371.1
Gene
ZNF627
Gene Name
zinc finger protein 627
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290083.1 824 Intron NP_001277012.1
NM_001290084.1 824 Intron NP_001277013.1
NM_001290085.1 824 Intron NP_001277014.1
NM_145295.3 824 Intron NP_660338.1
XM_011527780.2 824 Intron XP_011526082.1

View Full Product Details