Product Details

SNP ID
rs200788584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56441691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGGTTTCTCTTCAGAATGAATA[C/T]TCTGATGTATTTTAAGGTTTGCAGT
Phenotype
MIM: 611024
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF667 PubMed Links

Gene Details

Gene
ZNF667
Gene Name
zinc finger protein 667
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321355.1 1493 Missense Mutation AAT,AGT N378S NP_001308284.1
NM_001321356.1 1493 Missense Mutation AAT,AGT N435S NP_001308285.1
NM_022103.3 1493 Missense Mutation AAT,AGT N435S NP_071386.3
XM_011527208.1 1493 Missense Mutation AAT,AGT N435S XP_011525510.1
XM_011527209.1 1493 Missense Mutation AAT,AGT N435S XP_011525511.1

View Full Product Details