Product Details

SNP ID
rs200893734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9566847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGCTTCCTCACAGTCACTGTAT[G/T]CAAAGGATTTGTCTCCTATCCACGT
Phenotype
MIM: 194628
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF121 PubMed Links

Gene Details

Gene
ZNF121
Gene Name
zinc finger protein 121
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008727.3 498 Missense Mutation GAA,GCA E89A NP_001008727.1
NM_001308269.1 498 Missense Mutation GAA,GCA E89A NP_001295198.1
XM_017027239.1 498 Missense Mutation GAA,GCA E89A XP_016882728.1

View Full Product Details