Product Details

SNP ID
rs201235437
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57547118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTCCGGTGAAAGGCTTTCCCAC[A/G]CTGGGTGCATTCATAGGGCTTCTCC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ZNF549 PubMed Links

Gene Details

Gene
ZNF549
Gene Name
zinc finger protein 549
There are no transcripts associated with this gene.

Gene
ZNF550
Gene Name
zinc finger protein 550
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277090.1 1304 Missense Mutation CGT,TGT R376C NP_001264019.1
NM_001277091.1 1304 Missense Mutation CGT,TGT R376C NP_001264020.1
NM_001277092.1 1304 Missense Mutation CGT,TGT R376C NP_001264021.1
NM_001277093.1 1304 Intron NP_001264022.1
XM_011526567.2 1304 Missense Mutation CGT,TGT R376C XP_011524869.1
XM_011526568.2 1304 Missense Mutation CGT,TGT R367C XP_011524870.1
XM_017026401.1 1304 Missense Mutation CGT,TGT R376C XP_016881890.1

View Full Product Details