Product Details

SNP ID
rs201343885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11832726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTGGAGAGAAGCCCTATAAATG[C/T]GAGCAATGTGGGAAAGCCTTCAGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF440 PubMed Links

Gene Details

Gene
ZNF440
Gene Name
zinc finger protein 440
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152357.2 1584 Missense Mutation GCG,GTG A517V NP_689570.2
XM_005259731.3 1584 Missense Mutation GCG,GTG A520V XP_005259788.1
XM_017026253.1 1584 Missense Mutation GCG,GTG A519V XP_016881742.1
XM_017026254.1 1584 Missense Mutation GCG,GTG A395V XP_016881743.1

View Full Product Details