Product Details

SNP ID
rs202073531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50791683 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACATCCGCAGCACGGACTTTGAC[C/T]GCACGCTGGAGAGTGCCCAGGCCAA
Phenotype
MIM: 606362
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACPT PubMed Links

Gene Details

Gene
ACPT
Gene Name
acid phosphatase, testicular
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033068.2 331 Missense Mutation CGC,TGC R111C NP_149059.1
Gene
C19orf48
Gene Name
chromosome 19 open reading frame 48
There are no transcripts associated with this gene.

Gene
LOC105372440
Gene Name
uncharacterized LOC105372440
There are no transcripts associated with this gene.

Gene
SNORD88A
Gene Name
small nucleolar RNA, C/D box 88A
There are no transcripts associated with this gene.

Gene
SNORD88B
Gene Name
small nucleolar RNA, C/D box 88B
There are no transcripts associated with this gene.

View Full Product Details