Product Details

SNP ID
rs200655498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:29817268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTGCACCATCCAGAGGCTCCCC[A/G]CTGCCTGTACTGAGGTCAGTGCCGA
Phenotype
MIM: 123837
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCNE1 PubMed Links

Gene Details

Gene
CCNE1
Gene Name
cyclin E1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001238.3 495 Silent Mutation CCA,CCG P104P NP_001229.1
NM_001322259.1 495 Silent Mutation CCA,CCG P104P NP_001309188.1
NM_001322261.1 495 Silent Mutation CCA,CCG P104P NP_001309190.1
NM_001322262.1 495 Silent Mutation CCA,CCG P89P NP_001309191.1
XM_011527440.2 495 Silent Mutation CCA,CCG P101P XP_011525742.1

View Full Product Details