Product Details

SNP ID
rs200482859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18257304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCCAGGTGGCTTGAACTATCTTC[A/G]CTGCCATTTGCTGCTGCCTCATCTG
Phenotype
MIM: 600128
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA1683 PubMed Links

Gene Details

Gene
KIAA1683
Gene Name
KIAA1683
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145304.1 4196 Missense Mutation GCG,GTG A1327V NP_001138776.1
NM_001145305.1 4196 Missense Mutation GCG,GTG A1094V NP_001138777.1
NM_025249.3 4196 Missense Mutation GCG,GTG A1140V NP_079525.1
XM_005260084.1 4196 Missense Mutation GCG,GTG A1327V XP_005260141.1
XM_011528324.1 4196 Missense Mutation GCG,GTG A1281V XP_011526626.1
Gene
LOC729966
Gene Name
uncharacterized LOC729966
There are no transcripts associated with this gene.

Gene
PDE4C
Gene Name
phosphodiesterase 4C
There are no transcripts associated with this gene.

View Full Product Details