Product Details

SNP ID
rs200797935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58477325 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTTCCGAGGGTTCTGCTACCAGG[A/T]GGTGGCAGGTCCCCGAGAAGCCCTG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF324 PubMed Links

Gene Details

Gene
ZNF324
Gene Name
zinc finger protein 324
There are no transcripts associated with this gene.

Gene
ZNF446
Gene Name
zinc finger protein 446
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304453.1 488 Missense Mutation GAG,GTG E36V NP_001291382.1
NM_017908.3 488 Missense Mutation GAG,GTG E36V NP_060378.1
XM_005259052.4 488 Missense Mutation GAG,GTG E36V XP_005259109.1
XM_005259053.4 488 Missense Mutation GAG,GTG E36V XP_005259110.1
XM_006723266.3 488 Missense Mutation GAG,GTG E36V XP_006723329.1
XM_017026949.1 488 Missense Mutation GAG,GTG E36V XP_016882438.1
XM_017026950.1 488 Missense Mutation GAG,GTG E36V XP_016882439.1

View Full Product Details