Product Details

SNP ID
rs201349900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42078720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTGCTTATGCACCAAAACTCCCA[C/T]GTGCCCCAGCAGCACTTTGAGCTGG
Phenotype
MIM: 600283 MIM: 164176
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRIK5 PubMed Links

Gene Details

Gene
GRIK5
Gene Name
glutamate ionotropic receptor kainate type subunit 5
There are no transcripts associated with this gene.

Gene
POU2F2
Gene Name
POU class 2 homeobox 2
There are no transcripts associated with this gene.

Gene
ZNF574
Gene Name
zinc finger protein 574
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022752.5 146 Silent Mutation CAC,CAT H38H NP_073589.4
XM_005259159.2 146 Silent Mutation CAC,CAT H38H XP_005259216.1
XM_011527228.2 146 Silent Mutation CAC,CAT H38H XP_011525530.1
XM_011527229.2 146 Silent Mutation CAC,CAT H38H XP_011525531.1
XM_011527230.2 146 Silent Mutation CAC,CAT H38H XP_011525532.1
XM_017027149.1 146 Silent Mutation CAC,CAT H128H XP_016882638.1

View Full Product Details