Product Details

SNP ID
rs201591361
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54727752 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGGATCACTGAGGACCCCTTGC[G/T]CCTCGTTGGACAGCTCCACGATGCG
Phenotype
MIM: 610095
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIR3DL3 PubMed Links

Gene Details

Gene
KIR3DL3
Gene Name
killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153443.4 547 Missense Mutation CGC,CTC R166L NP_703144.3

View Full Product Details