Product Details

SNP ID
rs202186487
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1003179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCCAGCCCCCTGGAGACGCTGC[C/T]GGATGTGCTGGTGGCGGTGCTGCAG
Phenotype
MIM: 606651 MIM: 611011
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
GRIN3B PubMed Links
Additional Information
For this assay, SNP(s) [rs2240154] are located under a probe and SNP(s) [rs35592366] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
GRIN3B
Gene Name
glutamate ionotropic receptor NMDA type subunit 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138690.2 495 Missense Mutation CCG,CTG P159L NP_619635.1
XM_017026243.1 495 Intron XP_016881732.1
Gene
LOC105372235
Gene Name
uncharacterized LOC105372235
There are no transcripts associated with this gene.

Gene
TMEM259
Gene Name
transmembrane protein 259
There are no transcripts associated with this gene.

Gene
WDR18
Gene Name
WD repeat domain 18
There are no transcripts associated with this gene.

View Full Product Details