Product Details

SNP ID
rs200835105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40394222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGCCAGGCCTACACGTGGCAAG[C/T]GGACCCGGACCCGGCCCCGGCGACC
Phenotype
MIM: 611712 MIM: 605725
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIPK4 PubMed Links

Gene Details

Gene
HIPK4
Gene Name
homeodomain interacting protein kinase 4
There are no transcripts associated with this gene.

Gene
PRX
Gene Name
periaxin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020956.2 5048 UTR 3 NP_066007.1
NM_181882.2 5048 Missense Mutation CAC,CGC H1377R NP_870998.2
XM_011527171.2 5048 Missense Mutation CAC,CGC H1377R XP_011525473.1
XM_017027046.1 5048 Missense Mutation CAC,CGC H1343R XP_016882535.1
XM_017027047.1 5048 Missense Mutation CAC,CGC H1343R XP_016882536.1

View Full Product Details