Product Details

SNP ID
rs201695455
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6427398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGAGCATATTGGGCAGGTAGCCG[C/T]GGTAAAGGGCGCGGGTGCCCTCTCG
Phenotype
MIM: 603445 MIM: 608746 MIM: 610822
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KHSRP PubMed Links
Additional Information
For this assay, SNP(s) [rs3760750] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
KHSRP
Gene Name
KH-type splicing regulatory protein
There are no transcripts associated with this gene.

Gene
SLC25A23
Gene Name
solute carrier family 25 member 23
There are no transcripts associated with this gene.

Gene
SLC25A41
Gene Name
solute carrier family 25 member 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321298.1 608 Missense Mutation CAC,CGC H105R NP_001308227.1
NM_173637.3 608 Missense Mutation CAC,CGC H243R NP_775908.2
XM_011527926.1 608 Missense Mutation CAC,CGC H179R XP_011526228.1

View Full Product Details