Product Details

SNP ID
rs201696504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57280830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGGCGGCGGCGTGGATGGCTCC[C/G]GCGCAGGTGAGTGGACGAGGTTTCG
Phenotype
MIM: 604755
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC105372476 PubMed Links

Gene Details

Gene
LOC105372476
Gene Name
uncharacterized LOC105372476
There are no transcripts associated with this gene.

Gene
ZNF460
Gene Name
zinc finger protein 460
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006635.3 346 Silent Mutation CCC,CCG P8P NP_006626.3
XM_005258422.3 346 Intron XP_005258479.1
XM_017026176.1 346 Intron XP_016881665.1

View Full Product Details