Product Details

SNP ID
rs202083967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48635918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCCGGGGCATCCGCCGGGCCC[A/G]CAGTCTCCAGGCCTGGCCCAGGGGT
Phenotype
MIM: 604644 MIM: 124097 MIM: 607092
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CA11 PubMed Links

Gene Details

Gene
CA11
Gene Name
carbonic anhydrase 11
There are no transcripts associated with this gene.

Gene
DBP
Gene Name
D-box binding PAR bZIP transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001352.4 672 Missense Mutation GCG,GTG A71V NP_001343.2
XM_017026388.1 672 UTR 5 XP_016881877.1
Gene
SEC1P
Gene Name
secretory blood group 1, pseudogene
There are no transcripts associated with this gene.

Gene
SPHK2
Gene Name
sphingosine kinase 2
There are no transcripts associated with this gene.

View Full Product Details