Product Details

SNP ID
rs201493110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35896757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTATGCTCACGAATGTCAAGACGC[C/T]GGTACACCTGGAGCACCTCAGCCGC
Phenotype
MIM: 604089 MIM: 604142
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HCST PubMed Links

Gene Details

Gene
HCST
Gene Name
hematopoietic cell signal transducer
There are no transcripts associated with this gene.

Gene
NFKBID
Gene Name
NFKB inhibitor delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321831.1 545 Missense Mutation CAG,CGG Q91R NP_001308760.1
NM_139239.2 545 Missense Mutation CAG,CGG Q76R NP_640332.1
XM_005259340.3 545 Missense Mutation CAG,CGG Q91R XP_005259397.1
XM_011527419.2 545 Missense Mutation CAG,CGG Q228R XP_011525721.1
XM_011527420.2 545 Missense Mutation CAG,CGG Q91R XP_011525722.1
XM_011527421.2 545 Missense Mutation CAG,CGG Q66R XP_011525723.1
XM_011527424.2 545 Missense Mutation CAG,CGG Q91R XP_011525726.1
XM_017027389.1 545 Missense Mutation CAG,CGG Q91R XP_016882878.1
XM_017027390.1 545 Missense Mutation CAG,CGG Q91R XP_016882879.1
XM_017027391.1 545 Missense Mutation CAG,CGG Q76R XP_016882880.1
XM_017027392.1 545 Missense Mutation CAG,CGG Q76R XP_016882881.1
XM_017027393.1 545 Missense Mutation CAG,CGG Q76R XP_016882882.1
XM_017027394.1 545 Missense Mutation CAG,CGG Q66R XP_016882883.1
XM_017027395.1 545 Missense Mutation CAG,CGG Q66R XP_016882884.1
XM_017027396.1 545 Missense Mutation CAG,CGG Q76R XP_016882885.1
XM_017027397.1 545 Missense Mutation CAG,CGG Q43R XP_016882886.1
Gene
TYROBP
Gene Name
TYRO protein tyrosine kinase binding protein
There are no transcripts associated with this gene.

View Full Product Details