Product Details

SNP ID
rs207477287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57720550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTTTCCGGATGAATTCTCTCCC[G/T]CACTCACTACACACATAAGGCCTTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF671 PubMed Links

Gene Details

Gene
ZNF671
Gene Name
zinc finger protein 671
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321375.1 2128 Silent Mutation NP_001308304.1
NM_001321376.1 2128 Silent Mutation NP_001308305.1
NM_024833.2 2128 Silent Mutation NP_079109.2
XM_017027314.1 2128 Silent Mutation XP_016882803.1

View Full Product Details