Product Details

SNP ID
rs281860298
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11378276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGCTCCCTCTGGGCTGGGCTTC[T/G]AGGCCAAAGCAGATGAGCAGGAGGA
Phenotype
MIM: 133171 MIM: 614536
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
EPOR PubMed Links

Gene Details

Gene
EPOR
Gene Name
erythropoietin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000121.3 1370 Missense Mutation NP_000112.1
Gene
SWSAP1
Gene Name
SWIM-type zinc finger 7 associated protein 1
There are no transcripts associated with this gene.

View Full Product Details