Product Details

SNP ID
rs201426734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49583659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGGAACCTGGAACGGGAGTGTC[G/T]GGAAGAGAGGTGTTCCTGGGAAGAG
Phenotype
MIM: 616759 MIM: 616633 MIM: 604429
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NOSIP PubMed Links

Gene Details

Gene
NOSIP
Gene Name
nitric oxide synthase interacting protein
There are no transcripts associated with this gene.

Gene
PRR12
Gene Name
proline rich 12
There are no transcripts associated with this gene.

Gene
PRRG2
Gene Name
proline rich and Gla domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000951.2 535 Missense Mutation CGG,CTG R68L NP_000942.1
NM_001316335.1 535 Missense Mutation CGG,CTG R45L NP_001303264.1
XM_006723286.2 535 Missense Mutation CGG,CTG R68L XP_006723349.1
XM_011527124.1 535 Missense Mutation CGG,CTG R45L XP_011525426.1

View Full Product Details