Product Details

SNP ID
rs200566934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51029428 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACACCAGACCTTGAAGGACTCC[C/G]CCACACACCAGGGGGCCCCCAGAAT
Phenotype
MIM: 602673 MIM: 604434 MIM: 605539
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KLK10 PubMed Links

Gene Details

Gene
KLK10
Gene Name
kallikrein related peptidase 10
There are no transcripts associated with this gene.

Gene
KLK11
Gene Name
kallikrein related peptidase 11
There are no transcripts associated with this gene.

Gene
KLK12
Gene Name
kallikrein related peptidase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019598.2 478 Silent Mutation GGC,GGG G207G NP_062544.1
NM_145894.1 478 Silent Mutation GGC,GGG G207G NP_665901.1
NM_145895.1 478 UTR 3 NP_665902.1
XM_005258950.3 478 Silent Mutation GGC,GGG G97G XP_005259007.1
XM_005258951.3 478 UTR 3 XP_005259008.1

View Full Product Details