Product Details
- SNP ID
-
rs202089799
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57377197 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGTTTCTGTAGGAGTGTCACAGG[C/T]CATGGCTCCAAAGCCCTGTCTATCT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF547
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10413160] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF547
- Gene Name
- zinc finger protein 547
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173631.3 |
502 |
Missense Mutation |
GCC,GTC |
A74V |
NP_775902.2 |
View Full Product Details