Product Details

SNP ID
rs201361607
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7689363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCAGAAGGCGTCGTTCCAGCGAC[C/G]GGAGCCCCGCATCATCACGCAGTCC
Phenotype
MIM: 151445 MIM: 609565
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FCER2 PubMed Links

Gene Details

Gene
FCER2
Gene Name
Fc fragment of IgE receptor II
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207019.2 877 Missense Mutation CGT,GGT R265G NP_001193948.2
NM_001220500.1 877 Missense Mutation CGT,GGT R266G NP_001207429.1
NM_002002.4 877 Missense Mutation CGT,GGT R266G NP_001993.2
XM_005272462.4 877 Missense Mutation CGT,GGT R266G XP_005272519.1
Gene
MCEMP1
Gene Name
mast cell expressed membrane protein 1
There are no transcripts associated with this gene.

Gene
TRAPPC5
Gene Name
trafficking protein particle complex 5
There are no transcripts associated with this gene.

View Full Product Details