Product Details
- SNP ID
-
rs201774240
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57377294 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACATTCTGCGTCTGGCTGAGCATGA[C/T]GGAACACACCCCGAGCAGGGACTGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF547
PubMed Links
Gene Details
- Gene
- ZNF547
- Gene Name
- zinc finger protein 547
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173631.3 |
599 |
Silent Mutation |
GAC,GAT |
D106D |
NP_775902.2 |
View Full Product Details