Product Details

SNP ID
rs200914663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:20006993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTGCATACATTTGAAAAGTTTC[C/T]CTGTAGTATGTCTTATGTTTTCTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF682 PubMed Links

Gene Details

Gene
ZNF682
Gene Name
zinc finger protein 682
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077349.1 736 Intron NP_001070817.1
NM_033196.2 736 Intron NP_149973.1
XM_017027455.1 736 Missense Mutation GAG,GGG E176G XP_016882944.1
XM_017027456.1 736 Intron XP_016882945.1

View Full Product Details