Product Details

SNP ID
rs201533739
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45164201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAGATACTGCAGGCCAGAGGCC[A/G]TCGGGAGGAGGAGGGGGAAGCTGTT
Phenotype
MIM: 610396
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NKPD1 PubMed Links

Gene Details

Gene
NKPD1
Gene Name
NTPase, KAP family P-loop domain containing 1
There are no transcripts associated with this gene.

Gene
TRAPPC6A
Gene Name
trafficking protein particle complex 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270891.1 342 Missense Mutation NP_001257820.1
NM_001270892.1 342 Silent Mutation NP_001257821.1
NM_001270893.1 342 Silent Mutation NP_001257822.1
NM_024108.2 342 Missense Mutation NP_077013.1
XM_017027295.1 342 Silent Mutation XP_016882784.1

View Full Product Details