Product Details

SNP ID
rs202197333
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9413763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGGCGAGAGGATCTGGCAAAGG[C/T]CTTTCCACATTCCTTACATTCATAG
Phenotype
MIM: 604751
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF266 PubMed Links

Gene Details

Gene
ZNF266
Gene Name
zinc finger protein 266
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271314.1 2021 Missense Mutation NP_001258243.1
NM_006631.3 2021 Missense Mutation NP_006622.2
XM_005259716.3 2021 Missense Mutation XP_005259773.1
XM_006722620.2 2021 Missense Mutation XP_006722683.1
XM_006722621.2 2021 Missense Mutation XP_006722684.1
XM_006722622.2 2021 Missense Mutation XP_006722685.1
XM_006722623.3 2021 Missense Mutation XP_006722686.1
XM_006722627.2 2021 Missense Mutation XP_006722690.1
XM_006722630.2 2021 Missense Mutation XP_006722693.1
XM_011527644.1 2021 Missense Mutation XP_011525946.1
XM_011527647.1 2021 Missense Mutation XP_011525949.1
XM_011527648.1 2021 Missense Mutation XP_011525950.1
XM_011527650.1 2021 Missense Mutation XP_011525952.1
XM_017026154.1 2021 Missense Mutation XP_016881643.1
XM_017026155.1 2021 Missense Mutation XP_016881644.1
XM_017026156.1 2021 Missense Mutation XP_016881645.1
XM_017026157.1 2021 Missense Mutation XP_016881646.1
XM_017026158.1 2021 Missense Mutation XP_016881647.1
XM_017026159.1 2021 Missense Mutation XP_016881648.1
XM_017026160.1 2021 Missense Mutation XP_016881649.1
XM_017026161.1 2021 Missense Mutation XP_016881650.1
XM_017026162.1 2021 Missense Mutation XP_016881651.1
XM_017026163.1 2021 Missense Mutation XP_016881652.1
XM_017026164.1 2021 Missense Mutation XP_016881653.1
XM_017026165.1 2021 Missense Mutation XP_016881654.1
XM_017026166.1 2021 Missense Mutation XP_016881655.1
XM_017026167.1 2021 Missense Mutation XP_016881656.1
XM_017026168.1 2021 Missense Mutation XP_016881657.1
XM_017026169.1 2021 Missense Mutation XP_016881658.1
XM_017026170.1 2021 Missense Mutation XP_016881659.1
XM_017026171.1 2021 Missense Mutation XP_016881660.1
XM_017026172.1 2021 Missense Mutation XP_016881661.1
XM_017026173.1 2021 UTR 3 XP_016881662.1

View Full Product Details