Product Details

SNP ID
rs200606790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10320668 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCACTCACCGAGGGGTAGTCG[A/G]AGCTGTAGCTGTCAGACAGTCCTGG
Phenotype
MIM: 614585 MIM: 609950
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FDX1L PubMed Links

Gene Details

Gene
FDX1L
Gene Name
ferredoxin 1 like
There are no transcripts associated with this gene.

Gene
RAVER1
Gene Name
ribonucleoprotein, PTB binding 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133452.2 1888 Missense Mutation TCC,TTC S603F NP_597709.2
XM_011527671.1 1888 Missense Mutation TCC,TTC S578F XP_011525973.1

View Full Product Details