Product Details
- SNP ID
-
rs200813994
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:49480311 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTCTCCCCAGACTCCTCAACCCT[G/T]CCACCCCCATGGAGTCCCCGGCCCC
- Phenotype
-
MIM: 613358
MIM: 600007
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ALDH16A1
PubMed Links
Gene Details
- Gene
- ALDH16A1
- Gene Name
- aldehyde dehydrogenase 16 family member A1
There are no transcripts associated with this gene.
- Gene
- FLT3LG
- Gene Name
- fms related tyrosine kinase 3 ligand
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001204502.1 |
457 |
Silent Mutation |
CTG,CTT |
L165L |
NP_001191431.1 |
NM_001204503.1 |
457 |
Silent Mutation |
CTG,CTT |
L165L |
NP_001191432.1 |
NM_001278637.1 |
457 |
Silent Mutation |
CTG,CTT |
L83L |
NP_001265566.1 |
NM_001278638.1 |
457 |
Silent Mutation |
CTG,CTT |
L83L |
NP_001265567.1 |
NM_001459.3 |
457 |
Silent Mutation |
CTG,CTT |
L165L |
NP_001450.2 |
XM_005258680.4 |
457 |
Silent Mutation |
CTG,CTT |
L147L |
XP_005258737.3 |
XM_005258681.4 |
457 |
Silent Mutation |
CTG,CTT |
L147L |
XP_005258738.3 |
XM_005258682.4 |
457 |
Missense Mutation |
TGC,TTC |
C119F |
XP_005258739.3 |
XM_005258683.4 |
457 |
Missense Mutation |
TGC,TTC |
C119F |
XP_005258740.3 |
XM_006723116.3 |
457 |
Missense Mutation |
TGC,TTC |
C119F |
XP_006723179.2 |
XM_011526675.2 |
457 |
Intron |
|
|
XP_011524977.1 |
XM_011526676.2 |
457 |
Intron |
|
|
XP_011524978.1 |
XM_011526677.2 |
457 |
Intron |
|
|
XP_011524979.1 |
XM_011526678.2 |
457 |
Intron |
|
|
XP_011524980.1 |
XM_011526680.2 |
457 |
Intron |
|
|
XP_011524982.1 |
XM_011526682.2 |
457 |
Silent Mutation |
CTG,CTT |
L83L |
XP_011524984.1 |
XM_017026532.1 |
457 |
Missense Mutation |
TGC,TTC |
C119F |
XP_016882021.1 |
XM_017026533.1 |
457 |
Intron |
|
|
XP_016882022.1 |
XM_017026534.1 |
457 |
Intron |
|
|
XP_016882023.1 |
XM_017026535.1 |
457 |
Silent Mutation |
CTG,CTT |
L65L |
XP_016882024.1 |
- Gene
- RPL13A
- Gene Name
- ribosomal protein L13a
There are no transcripts associated with this gene.
- Gene
- SNORD32A
- Gene Name
- small nucleolar RNA, C/D box 32A
There are no transcripts associated with this gene.
View Full Product Details