Product Details

SNP ID
rs201019332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:58477233 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGAGCAAGAATGCCATCCCCTCT[A/G]GGTCCCCCATGCCTGCCCGTCATGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF324 PubMed Links

Gene Details

Gene
ZNF324
Gene Name
zinc finger protein 324
There are no transcripts associated with this gene.

Gene
ZNF446
Gene Name
zinc finger protein 446
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304453.1 396 Silent Mutation CTA,CTG L5L NP_001291382.1
NM_017908.3 396 Silent Mutation CTA,CTG L5L NP_060378.1
XM_005259052.4 396 Silent Mutation CTA,CTG L5L XP_005259109.1
XM_005259053.4 396 Silent Mutation CTA,CTG L5L XP_005259110.1
XM_006723266.3 396 Silent Mutation CTA,CTG L5L XP_006723329.1
XM_017026949.1 396 Silent Mutation CTA,CTG L5L XP_016882438.1
XM_017026950.1 396 Silent Mutation CTA,CTG L5L XP_016882439.1

View Full Product Details