Product Details

SNP ID
rs201295756
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57720598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCCAGTGTGGACTTTCCAGTG[C/T]CTGATGAGGTTGGGTCTTTGAGTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF671 PubMed Links

Gene Details

Gene
ZNF671
Gene Name
zinc finger protein 671
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321375.1 2080 Silent Mutation AGA,AGG R398R NP_001308304.1
NM_001321376.1 2080 Silent Mutation AGA,AGG R419R NP_001308305.1
NM_024833.2 2080 Silent Mutation AGA,AGG R496R NP_079109.2
XM_017027314.1 2080 Silent Mutation AGA,AGG R419R XP_016882803.1

View Full Product Details