Product Details

SNP ID
rs201469472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2477137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCCAGTCCTTCTGCTGTGACAA[C/G]GACATCAACATCGTGCGGGTGTCGG
Phenotype
MIM: 604948
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GADD45B PubMed Links

Gene Details

Gene
GADD45B
Gene Name
growth arrest and DNA damage inducible beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015675.3 489 Missense Mutation AAC,AAG N85K NP_056490.2
XM_017026822.1 489 Missense Mutation AAC,AAG N85K XP_016882311.1

View Full Product Details