Product Details

SNP ID
hCV190527188
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:2463898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAAGTGGAACTCGAGCAATACCA[G/A]TCTGAAAAATAAACAAATATGCTCT
Phenotype
MIM: 182282
Polymorphism
G/A, Transition Substitution
Allele Nomenclature
Literature Links
SNORD119 PubMed Links

Gene Details

Gene
SNORD119
Gene Name
small nucleolar RNA, C/D box 119
There are no transcripts associated with this gene.

Gene
SNRPB
Gene Name
small nuclear ribonucleoprotein polypeptides B and B1
There are no transcripts associated with this gene.

View Full Product Details