Product Details

SNP ID
rs201230566
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:109606018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATATCATAGGCACTGAGGGCTGC[A/G]CAGAAAATGATGCAGGTGACTCCCT
Phenotype
MIM: 139340 MIM: 611189
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNAT2 PubMed Links

Gene Details

Gene
GNAT2
Gene Name
G protein subunit alpha transducin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005272.3 885 Silent Mutation TGC,TGT C224C NP_005263.1
XM_011541264.2 885 Silent Mutation TGC,TGT C224C XP_011539566.1
Gene
MIR197
Gene Name
microRNA 197
There are no transcripts associated with this gene.

View Full Product Details