Product Details

SNP ID
rs201321708
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:152972057 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAGGTCAAGAAGCAATGCCCAC[C/T]GAAAGGCACCATCATTCCAGCCCAG
Phenotype
MIM: 616363
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPRR4 PubMed Links

Gene Details

Gene
SPRR4
Gene Name
small proline rich protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173080.2 925 Missense Mutation CCG,CTG P56L NP_775103.1
XM_017000482.1 925 Missense Mutation CCG,CTG P56L XP_016855971.1

View Full Product Details