Product Details

SNP ID
rs201625211
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:241500443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTCCTTAAACACTTACCTATAT[A/G]AGGATTGAGAGCTGTCACCAACATT
Phenotype
MIM: 136850
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FH PubMed Links

Gene Details

Gene
FH
Gene Name
fumarate hydratase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000143.3 1887 Missense Mutation CAT,TAT H462Y NP_000134.2
XM_011544132.2 1887 Missense Mutation CAT,TAT H386Y XP_011542434.1

View Full Product Details